Product Details

SNP ID
rs201429667
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99533276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTCTGCGCCCTGCATGCTGGCC[A/G]CCGCTGAGGGGACGCAATTTTCTGA
Phenotype
MIM: 606727
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC01475 PubMed Links
Additional Information
For this assay, SNP(s) [rs41290504] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LINC01475
Gene Name
long intergenic non-protein coding RNA 1475
There are no transcripts associated with this gene.

Gene
NKX2-3
Gene Name
NK2 homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145285.2 344 Missense Mutation ACC,GCC T49A NP_660328.2
XM_011539370.1 344 Missense Mutation ACC,GCC T49A XP_011537672.1

View Full Product Details