Product Details

SNP ID
rs199554891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:83165797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTAACCGGGATCCTCGTCTGAA[C/T]AGGATAAGCCAACATTCTCATGGAA
Phenotype
MIM: 608876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCF11 PubMed Links

Gene Details

Gene
PCF11
Gene Name
PCF11 cleavage and polyadenylation factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015885.3 1245 Silent Mutation AAC,AAT N300N NP_056969.2
XM_005274048.1 1245 Silent Mutation AAC,AAT N300N XP_005274105.1
XM_005274049.1 1245 Silent Mutation AAC,AAT N300N XP_005274106.1
XM_017017918.1 1245 Silent Mutation AAC,AAT N300N XP_016873407.1

View Full Product Details