Product Details

SNP ID
rs199504970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123058705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGCAGAGACATTGAGTATACC[A/G]TTGGCATCAATGTCAAAAGTGACTT
Phenotype
MIM: 600816
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPA8 PubMed Links

Gene Details

Gene
HSPA8
Gene Name
heat shock protein family A (Hsp70) member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006597.5 1726 Silent Mutation AAC,AAT N483N NP_006588.1
NM_153201.3 1726 Intron NP_694881.1
XM_011542798.1 1726 Intron XP_011541100.1
Gene
SNORD14C
Gene Name
small nucleolar RNA, C/D box 14C
There are no transcripts associated with this gene.

Gene
SNORD14D
Gene Name
small nucleolar RNA, C/D box 14D
There are no transcripts associated with this gene.

Gene
SNORD14E
Gene Name
small nucleolar RNA, C/D box 14E
There are no transcripts associated with this gene.

View Full Product Details