Product Details

SNP ID
rs201014647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8937748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCAGGTAGTTAATGTACTTGAT[C/G]GCAGCTCTGAGGGTTTCCACTTTGC
Phenotype
MIM: 609154
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASCL3 PubMed Links

Gene Details

Gene
ASCL3
Gene Name
achaete-scute family bHLH transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020646.2 474 Silent Mutation GCC,GCG A138A NP_065697.1
Gene
C11orf16
Gene Name
chromosome 11 open reading frame 16
There are no transcripts associated with this gene.

Gene
TMEM9B
Gene Name
TMEM9 domain family member B
There are no transcripts associated with this gene.

View Full Product Details