Product Details

SNP ID
rs200882575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5736690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACACCCTCATCCTCATCATCAT[C/G]TGGCAGAACCCTTCTTTACAGCAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR52N4 PubMed Links

Gene Details

Gene
OR52N4
Gene Name
olfactory receptor family 52 subfamily N member 4 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005175.3 243 Intron NP_001005175.3
XM_017017711.1 243 Intron XP_016873200.1
XM_017017712.1 243 Intron XP_016873201.1
XM_017017713.1 243 Intron XP_016873202.1
Gene
OR56B1
Gene Name
olfactory receptor family 56 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005180.2 243 Missense Mutation ATC,ATG I58M NP_001005180.1

View Full Product Details