Product Details
- SNP ID
-
rs200974220
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:102864167 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACTGTTTTTAGAGTAGAAGACTG[A/C]ATCAATTTTAGGCCCGATTCCTTGG
- Phenotype
-
MIM: 601046
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MMP12
PubMed Links
Gene Details
- Gene
- MMP12
- Gene Name
- matrix metallopeptidase 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002426.5 |
1388 |
Missense Mutation |
GCA,TCA |
A431S |
NP_002417.2 |
View Full Product Details