Product Details

SNP ID
rs200974716
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94544859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGCTTTTCCACCACCGCGACCT[A/C]GTGAAGGGGCCCCCCGACTGGCCCC
Phenotype
MIM: 104230
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
FUT4 PubMed Links

Gene Details

Gene
FUT4
Gene Name
fucosyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002033.3 1009 Silent Mutation CTA,CTC L242L NP_002024.1
Gene
LOC105369438
Gene Name
uncharacterized LOC105369438
There are no transcripts associated with this gene.

View Full Product Details