Product Details

SNP ID
rs200751482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4914934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGATGTGGGAAACACAGGTGT[C/T]AAGGGCCTTGAATCTCTCAGCCCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51A7 PubMed Links

Gene Details

Gene
OR51A7
Gene Name
olfactory receptor family 51 subfamily A member 7
There are no transcripts associated with this gene.

Gene
OR51G1
Gene Name
olfactory receptor family 51 subfamily G member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR51G2
Gene Name
olfactory receptor family 51 subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005238.1 730 Missense Mutation AAC,GAC N244D NP_001005238.1

View Full Product Details