Product Details

SNP ID
rs201027576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117430734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGCACTCACTCTTGTTGGCCC[A/G]GTGAGGGATGGGCACAGCCACGTTT
Phenotype
MIM: 611782
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSCAML1 PubMed Links

Gene Details

Gene
DSCAML1
Gene Name
DS cell adhesion molecule like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020693.3 4133 Missense Mutation CGG,TGG R1952W NP_065744.2
XM_011542917.1 4133 Missense Mutation CGG,TGG R1904W XP_011541219.1
XM_011542918.1 4133 Missense Mutation CGG,TGG R1816W XP_011541220.1
XM_011542919.2 4133 Missense Mutation CGG,TGG R1465W XP_011541221.1
XM_011542920.2 4133 Missense Mutation CGG,TGG R1382W XP_011541222.1
XM_011542921.2 4133 Missense Mutation CGG,TGG R1183W XP_011541223.1
XM_011542924.2 4133 Missense Mutation CGG,TGG R1109W XP_011541226.1
XM_011542925.2 4133 Missense Mutation CGG,TGG R1103W XP_011541227.1

View Full Product Details