Product Details

SNP ID
rs201030643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73677905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGATGACATGGGAGATTAGCAG[A/G]AACAGCCTCCTTCACTGACTGGTTG
Phenotype
MIM: 179513
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB6A PubMed Links

Gene Details

Gene
RAB6A
Gene Name
RAB6A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243718.1 Intron NP_001230647.1
NM_001243719.1 Intron NP_001230648.1
NM_002869.4 Intron NP_002860.2
NM_198896.1 Intron NP_942599.1

View Full Product Details