Product Details
- SNP ID
-
rs200899138
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:7796294 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCATCTTCAGGATGGTGATGAGGA[C/T]ATAGATGTAGCAGACGGCTATGACA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR5P2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76615008] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR5P2
- Gene Name
- olfactory receptor family 5 subfamily P member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153444.1 |
649 |
Missense Mutation |
ATC,GTC |
I217V |
NP_703145.1 |
View Full Product Details