Product Details

SNP ID
rs200967365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10755773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTTTGTACTCAATCAGTCTCC[A/G]AATAATATTCCAGCCCTTCTTGGTA
Phenotype
MIM: 609366
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTR9 PubMed Links

Gene Details

Gene
CTR9
Gene Name
CTR9 homolog, Paf1/RNA polymerase II complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014633.4 906 Silent Mutation CCA,CCG P160P NP_055448.1

View Full Product Details