Product Details

SNP ID
rs200988172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57528834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCAGGTACTTAGAGACACATCG[C/G]TCCAGGCACACAGACTCGCCCTTGG
Phenotype
MIM: 613664 MIM: 602251
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMTNL1 PubMed Links

Gene Details

Gene
SMTNL1
Gene Name
smoothelin like 1
There are no transcripts associated with this gene.

Gene
TIMM10
Gene Name
translocase of inner mitochondrial membrane 10 homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012456.2 303 Missense Mutation GAC,GAG D52E NP_036588.1

View Full Product Details