Product Details

SNP ID
rs200856208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47216993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGTGGCTGTGGGTTCCAAAGGG[A/G]GAGATATCATGCTCTGGAATTTTGG
Phenotype
MIM: 600811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDB2 PubMed Links

Gene Details

Gene
DDB2
Gene Name
damage specific DNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000107.2 595 Missense Mutation AGA,GGA R134G NP_000098.1
NM_001300734.1 595 Missense Mutation AGA,GGA R134G NP_001287663.1

View Full Product Details