Product Details

SNP ID
rs200717826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5857181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAGGGATGATACGATGCCCACA[G/T]AAGGGCAGCCTCAGAAGGAGAAACA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR52E8 PubMed Links

Gene Details

Gene
OR52E8
Gene Name
olfactory receptor family 52 subfamily E member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005168.1 522 Missense Mutation TTA,TTC L174F NP_001005168.1

View Full Product Details