Product Details

SNP ID
rs200903478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102579135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTCCATTTTCCTTTTCCTTT[C/T]GTCGTAGCTAGAAAAAGTATTATTT
Phenotype
MIM: 604629
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP20 PubMed Links

Gene Details

Gene
MMP20
Gene Name
matrix metallopeptidase 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004771.3 1268 Missense Mutation AAA,GAA K419E NP_004762.2

View Full Product Details