Product Details

SNP ID
rs200087571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65641408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCGGACCTGCTGCATGGGGCA[A/C]CTGGCTTTGTGTGTGAGCTCGGGGG
Phenotype
MIM: 602180
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR4489 PubMed Links

Gene Details

Gene
MIR4489
Gene Name
microRNA 4489
There are no transcripts associated with this gene.

Gene
MIR4690
Gene Name
microRNA 4690
There are no transcripts associated with this gene.

Gene
PCNX3
Gene Name
pecanex homolog 3 (Drosophila)
There are no transcripts associated with this gene.

Gene
SIPA1
Gene Name
signal-induced proliferation-associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006747.3 694 Missense Mutation ACT,CCT T163P NP_006738.3
NM_153253.29 694 Missense Mutation ACT,CCT T163P NP_694985.29
XM_005274189.2 694 Missense Mutation ACT,CCT T163P XP_005274246.1
XM_011545214.1 694 Missense Mutation ACT,CCT T163P XP_011543516.1

View Full Product Details