Product Details

SNP ID
rs200003759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13010333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCTCCACGAGCTGGACCGCGAG[A/G]TCGATCACTACGAGGCCAAGGTGCA
Phenotype
MIM: 614713
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASSF10 PubMed Links

Gene Details

Gene
RASSF10
Gene Name
Ras association domain family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080521.2 911 Missense Mutation ATC,GTC I253V NP_001073990.2

View Full Product Details