Product Details

SNP ID
rs200254653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:555075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGTGTTCTGCACCGACAGGGCCA[C/G]CAGGGGGCAGCTACGGTCCACGCTC
Phenotype
MIM: 143023
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LMNTD2 PubMed Links

Gene Details

Gene
LMNTD2
Gene Name
lamin tail domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173573.2 1934 Missense Mutation CTG,GTG L604V NP_775844.2
XM_011519965.2 1934 Missense Mutation CTG,GTG L677V XP_011518267.2
XM_011519967.2 1934 Missense Mutation CTG,GTG L637V XP_011518269.2
XM_017017476.1 1934 Missense Mutation CTG,GTG L692V XP_016872965.1
XM_017017477.1 1934 Missense Mutation CTG,GTG L687V XP_016872966.1
XM_017017478.1 1934 Missense Mutation CTG,GTG L624V XP_016872967.1
XM_017017479.1 1934 Intron XP_016872968.1
Gene
LRRC56
Gene Name
leucine rich repeat containing 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198075.3 1934 Intron NP_932341.1
XM_005252775.2 1934 Intron XP_005252832.1
XM_005252776.2 1934 Intron XP_005252833.1
XM_006718132.2 1934 Intron XP_006718195.1
XM_006718133.2 1934 Intron XP_006718196.1
XM_011519875.2 1934 Intron XP_011518177.1
XM_011519876.1 1934 Intron XP_011518178.1
XM_011519877.2 1934 Intron XP_011518179.1
XM_017017167.1 1934 Intron XP_016872656.1
XM_017017168.1 1934 Intron XP_016872657.1
Gene
RASSF7
Gene Name
Ras association domain family member 7
There are no transcripts associated with this gene.

View Full Product Details