Product Details
- SNP ID
-
rs199983925
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:114443909 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAGTTGCTGGACAGCATGTCAGA[C/T]TGGTGTAAGGAGCATCACGGGAAGG
- Phenotype
-
MIM: 607149
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
REXO2
PubMed Links
Gene Details
- Gene
- REXO2
- Gene Name
- RNA exonuclease 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015523.3 |
428 |
Silent Mutation |
GAC,GAT |
D95D |
NP_056338.2 |
View Full Product Details