Product Details

SNP ID
rs199984646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63914310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTCTCCCTCACTCACGCCTCCT[C/T]GACCCTCTTCGAGCTCATCACTGCT
Phenotype
MIM: 600526 MIM: 616019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MARK2 PubMed Links

Gene Details

Gene
MARK2
Gene Name
microtubule affinity regulating kinase 2
There are no transcripts associated with this gene.

Gene
RCOR2
Gene Name
REST corepressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173587.3 1014 Missense Mutation CAA,CGA Q209R NP_775858.2
XM_005273932.3 1014 Intron XP_005273989.1

View Full Product Details