Product Details

SNP ID
rs200307365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68760329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTGTTGATAATCTCCAAGGCT[A/C]AGATAAAACCTATTGAGTGAAACAG
Phenotype
MIM: 600528 MIM: 604374
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPT1A PubMed Links

Gene Details

Gene
CPT1A
Gene Name
carnitine palmitoyltransferase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031847.2 2208 Nonsense Mutation GAG,TAG E680* NP_001027017.1
NM_001876.3 2208 Nonsense Mutation GAG,TAG E680* NP_001867.2
XM_005273762.2 2208 Nonsense Mutation GAG,TAG E712* XP_005273819.1
XM_005273763.1 2208 Nonsense Mutation GAG,TAG E712* XP_005273820.1
XM_017017220.1 2208 Nonsense Mutation GAG,TAG E680* XP_016872709.1
Gene
TESMIN
Gene Name
testis expressed metallothionein like protein
There are no transcripts associated with this gene.

View Full Product Details