Product Details

SNP ID
rs200657045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66482258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCGGCGTGTCTAGCCTGGACTGC[C/T]GTGAGGCCTTCCGCCTGCTGTCACC
Phenotype
MIM: 606818 MIM: 609827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPP3 PubMed Links

Gene Details

Gene
DPP3
Gene Name
dipeptidyl peptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256670.1 519 Missense Mutation CGT,TGT R20C NP_001243599.1
NM_005700.4 519 Missense Mutation CGT,TGT R20C NP_005691.2
NM_130443.3 519 Missense Mutation CGT,TGT R20C NP_569710.2
Gene
LOC101928069
Gene Name
uncharacterized LOC101928069
There are no transcripts associated with this gene.

Gene
PELI3
Gene Name
pellino E3 ubiquitin protein ligase family member 3
There are no transcripts associated with this gene.

View Full Product Details