Product Details

SNP ID
rs200342430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:105139066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGATAAAAATTCTTCTCTTTTTA[C/T]GCAAGAGTTGGTCTGTTGGAAGCAC
Phenotype
MIM: 605354
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD18 PubMed Links

Gene Details

Gene
CARD18
Gene Name
caspase recruitment domain family member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021571.3 28 Missense Mutation CAT,CGT H7R NP_067546.1

View Full Product Details