Product Details

SNP ID
rs200492555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59210883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAGAATGGTGGTGACCCCCAC[A/T]GTGACCCCAGCTGCAGCCCCTCCTG
Phenotype
MIM: 616110 MIM: 610390
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DTX4 PubMed Links

Gene Details

Gene
DTX4
Gene Name
deltex E3 ubiquitin ligase 4
There are no transcripts associated with this gene.

Gene
MPEG1
Gene Name
macrophage expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039396.1 Intron NP_001034485.1

View Full Product Details