Product Details

SNP ID
rs199946571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7796241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGAGGTGGGAAGTGCAGGTGGA[A/G]AAGGCCTTGTGGTGCCCCTCAGTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5P2 PubMed Links

Gene Details

Gene
OR5P2
Gene Name
olfactory receptor family 5 subfamily P member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153444.1 702 Silent Mutation TTC,TTT F234F NP_703145.1

View Full Product Details