Product Details

SNP ID
rs199740628
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.11:56417848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATCAGTGTTGAATAATGCAGGG[A/G]ACTACAGATGGCGACATAGCAATCG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR5R1 PubMed Links

Gene Details

Gene
OR5R1
Gene Name
olfactory receptor family 5 subfamily R member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004744.1 385 Missense Mutation CCC,TCC P129S NP_001004744.1

View Full Product Details