Product Details
- SNP ID
-
rs199877978
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:124613035 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTCAGCATCCGGCAGGCAGCCTA[C/T]GTGGACAGCTCCTGCTGGGACTCAC
- Phenotype
-
MIM: 608422
MIM: 610614
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PANX3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34498516] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PANX3
- Gene Name
- pannexin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052959.2 |
237 |
Silent Mutation |
TAC,TAT |
Y79Y |
NP_443191.1 |
- Gene
- TBRG1
- Gene Name
- transforming growth factor beta regulator 1
There are no transcripts associated with this gene.
View Full Product Details