Product Details

SNP ID
rs199627331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18137956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGATTGGAAAGTCTTATTTTGT[C/T]ATGTGCATCTAGTTTCCATTTTCCT
Phenotype
MIM: 607229
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRGPRX3 PubMed Links

Gene Details

Gene
MRGPRX3
Gene Name
MAS related GPR family member X3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054031.3 970 Intron NP_473372.3
XM_011519882.2 970 Missense Mutation CAT,TAT H252Y XP_011518184.1

View Full Product Details