Product Details

SNP ID
rs199721170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58837351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAATTTGTCAGGAGCTTTGGTG[A/G]AGATGTGGTAAGTGTTGGTATAATG
Phenotype
MIM: 614762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLYATL2 PubMed Links

Gene Details

Gene
GLYATL2
Gene Name
glycine-N-acyltransferase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145016.3 549 Missense Mutation TCC,TTC S78F NP_659453.3
XM_011544809.2 549 Missense Mutation TCC,TTC S37F XP_011543111.1
XM_017017337.1 549 Missense Mutation TCC,TTC S78F XP_016872826.1
XM_017017338.1 549 Missense Mutation TCC,TTC S78F XP_016872827.1

View Full Product Details