Product Details

SNP ID
rs199635205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124626917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTTTATAGATCATCACCGACC[A/G]ACCTGGCTTTCATGATGAGAGTGCC
Phenotype
MIM: 608422 MIM: 610079 MIM: 610614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PANX3 PubMed Links

Gene Details

Gene
PANX3
Gene Name
pannexin 3
There are no transcripts associated with this gene.

Gene
SIAE
Gene Name
sialic acid acetylesterase
There are no transcripts associated with this gene.

Gene
TBRG1
Gene Name
transforming growth factor beta regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032811.2 649 Missense Mutation CAA,CGA Q202R NP_116200.2
XM_005271709.3 649 Missense Mutation CAA,CGA Q55R XP_005271766.1
XM_017018442.1 649 Missense Mutation CAA,CGA Q51R XP_016873931.1

View Full Product Details