Product Details

SNP ID
rs199666697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58190656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATCCGTGGCGATCGTCGGCTC[C/T]ACACCCCGATGTACTTCTTCCTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR9Q1 PubMed Links

Gene Details

Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
There are no transcripts associated with this gene.

Gene
OR9Q2
Gene Name
olfactory receptor family 9 subfamily Q member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005283.2 223 Missense Mutation CAC,TAC H56Y NP_001005283.1

View Full Product Details