Product Details

SNP ID
rs199760382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130406119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGGGATGGTGACAATGTCATT[G/T]TAGCCATAACTGTGATAGAAACAGA
Phenotype
MIM: 605175
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS8 PubMed Links

Gene Details

Gene
ADAMTS8
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007037.5 2816 Nonsense Mutation TAA,TAC *703Y NP_008968.4
XM_017017145.1 2816 Nonsense Mutation TAA,TAC *687Y XP_016872634.1
Gene
LOC646383
Gene Name
uncharacterized LOC646383
There are no transcripts associated with this gene.

View Full Product Details