Product Details

SNP ID
rs201476861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67451748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTGGGTCTGTGGCTCAGTGGGC[A/G]TGAAGCTGCCCGGCCGCTCCTCGCA
Phenotype
MIM: 608965 MIM: 609849
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CABP4 PubMed Links

Gene Details

Gene
CABP4
Gene Name
calcium binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300895.1 982 Intron NP_001287824.1
NM_001300896.1 982 Intron NP_001287825.1
NM_145200.3 982 Intron NP_660201.1
XM_005274114.3 982 Intron XP_005274171.2
XM_011545181.2 982 Intron XP_011543483.1
XM_011545182.2 982 Intron XP_011543484.1
XM_011545183.2 982 Intron XP_011543485.1
XM_017018025.1 982 Intron XP_016873514.1
Gene
CORO1B
Gene Name
coronin 1B
There are no transcripts associated with this gene.

Gene
GPR152
Gene Name
G protein-coupled receptor 152
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206997.1 982 Missense Mutation ACG,ATG T326M NP_996880.1

View Full Product Details