Product Details

SNP ID
rs201525236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4385318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGGATCCTTGTGATCCAATATT[C/T]AGTGGACAGAGGGTTAGAGGGGCTG
Phenotype
MIM: 109092
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52B3P PubMed Links

Gene Details

Gene
OR52B3P
Gene Name
olfactory receptor family 52 subfamily B member 3 pseudogene
There are no transcripts associated with this gene.

Gene
TRIM21
Gene Name
tripartite motif containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003141.3 1508 Silent Mutation CTA,CTG L465L NP_003132.2

View Full Product Details