Product Details

SNP ID
rs201455616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57598295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCCTCCAGGCTGACCCTGCTG[A/C]CCCTCCTGCTGCTGCTGCTGGCTGG
Phenotype
MIM: 606860
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SERPING1 PubMed Links

Gene Details

Gene
SERPING1
Gene Name
serpin family G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000062.2 64 Missense Mutation ACC,CCC T9P NP_000053.2
NM_001032295.1 64 Missense Mutation ACC,CCC T9P NP_001027466.1

View Full Product Details