Product Details

SNP ID
rs201594600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65389495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTTCCAGACATCGCCCTGGAT[A/G]TCTTCGCGCTCTGGCTGGTCTCCCC
Phenotype
MIM: 610825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRMD8 PubMed Links

Gene Details

Gene
FRMD8
Gene Name
FERM domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300832.1 Intron NP_001287761.1
NM_001300833.1 Intron NP_001287762.1
NM_031904.3 Intron NP_114110.1
Gene
SLC25A45
Gene Name
solute carrier family 25 member 45
There are no transcripts associated with this gene.

View Full Product Details