Product Details

SNP ID
rs201621404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34883403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGACGTCTGACCAGTACTGCA[C/T]AGGAGTCTGGGTATTCATTCATTGC
Phenotype
MIM: 612491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APIP PubMed Links

Gene Details

Gene
APIP
Gene Name
APAF1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015957.3 690 Missense Mutation TAT,TGT Y188C NP_057041.2
XM_011520154.2 690 Intron XP_011518456.1
XM_017017875.1 690 Intron XP_016873364.1

View Full Product Details