Product Details

SNP ID
rs201752677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118349785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTATGACTATCAAGAAGATGGTT[C/T]GGTACTTCTGACTTGTGATGCAGAA
Phenotype
MIM: 186790 MIM: 186740 MIM: 603753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD3D PubMed Links

Gene Details

Gene
CD3D
Gene Name
CD3d molecule
There are no transcripts associated with this gene.

Gene
CD3G
Gene Name
CD3g molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000073.2 202 Missense Mutation TCG,TTG S41L NP_000064.1
XM_005271724.3 202 Missense Mutation TCG,TTG S41L XP_005271781.1
XM_006718941.2 202 Missense Mutation TCG,TTG S41L XP_006719004.1
Gene
UBE4A
Gene Name
ubiquitination factor E4A
There are no transcripts associated with this gene.

View Full Product Details