Product Details
- SNP ID
-
rs201705374
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:7089131 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCGACCGGCAAACATGGTTGAAG[C/G]GGATCGCCCGGGGAAGCTGTTCATT
- Phenotype
-
MIM: 609665
MIM: 605444
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NLRP14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4758158] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NLRP14
- Gene Name
- NLR family pyrin domain containing 14
- Gene
- RBMXL2
- Gene Name
- RNA binding motif protein, X-linked like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014469.4 |
198 |
Missense Mutation |
GCG,GGG |
A4G |
NP_055284.3 |
View Full Product Details