Product Details

SNP ID
rs201451089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6478122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGTGTTGATGCTAGAGACAAAG[A/T]AGTTCACGGCTCCTAGCAGCGTTTC
Phenotype
MIM: 601638 MIM: 607388 MIM: 605493
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARFIP2 PubMed Links

Gene Details

Gene
ARFIP2
Gene Name
ADP ribosylation factor interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242854.1 865 Missense Mutation TAC,TTC Y238F NP_001229783.1
NM_001242855.1 865 Missense Mutation TAC,TTC Y167F NP_001229784.1
NM_001242856.1 865 Missense Mutation TAC,TTC Y120F NP_001229785.1
NM_012402.3 865 Missense Mutation TAC,TTC Y205F NP_036534.1
XM_005252840.4 865 Missense Mutation TAC,TTC Y205F XP_005252897.1
Gene
TIMM10B
Gene Name
translocase of inner mitochondrial membrane 10 homolog B (yeast)
There are no transcripts associated with this gene.

Gene
TRIM3
Gene Name
tripartite motif containing 3
There are no transcripts associated with this gene.

View Full Product Details