Product Details

SNP ID
rs201729955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74267279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGGGCTGGAGCTGCAGGGTCTG[C/T]GGAATTCCTGTCCATACTCATGTAT
Phenotype
MIM: 608987
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P4HA3 PubMed Links

Gene Details

Gene
P4HA3
Gene Name
prolyl 4-hydroxylase subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288748.1 1669 Missense Mutation ACA,GCA T533A NP_001275677.1
NM_182904.4 1669 Missense Mutation CAC,CGC H535R NP_878907.1

View Full Product Details