Product Details

SNP ID
rs202172162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85685489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGTGCAGGCCTGCTGGCTCTG[C/T]AGCTCCAGCAGCTCCTTCTCGCGCG
Phenotype
MIM: 612880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC89 PubMed Links

Gene Details

Gene
CCDC89
Gene Name
coiled-coil domain containing 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152723.2 789 Silent Mutation CTA,CTG L214L NP_689936.1
Gene
SYTL2
Gene Name
synaptotagmin like 2
There are no transcripts associated with this gene.

View Full Product Details