Product Details

SNP ID
rs202156177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59576586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGTTACTGCCGTTTCAGAAAAT[G/T]TCCGGGCATGAGCTCCAGTCCTGTT
Phenotype
MIM: 167040
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OSBP PubMed Links

Gene Details

Gene
OSBP
Gene Name
oxysterol binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002556.2 2895 Silent Mutation NP_002547.1

View Full Product Details