Product Details

SNP ID
rs201976014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7488132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTATTTTCTGACTGAAGCAAGG[A/G]CTGGAAAAATGTACCCAAGCAACGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFML1 PubMed Links

Gene Details

Gene
OLFML1
Gene Name
olfactomedin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198474.3 642 Silent Mutation GGA,GGG G45G NP_940876.2
XM_005252878.4 642 Silent Mutation GGA,GGG G45G XP_005252935.1

View Full Product Details