Product Details

SNP ID
rs202160971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108477646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGTAGCAGTAAAGCCTGTGTGG[C/T]TGTAGTAGGTCCCTAGCCATCAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf65 PubMed Links

Gene Details

Gene
C11orf65
Gene Name
chromosome 11 open reading frame 65
There are no transcripts associated with this gene.

Gene
KDELC2
Gene Name
KDEL motif containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153705.4 1398 Silent Mutation CAA,CAG Q453Q NP_714916.3
XM_011542621.2 1398 Silent Mutation CAA,CAG Q439Q XP_011540923.1
XM_011542623.2 1398 Silent Mutation CAA,CAG Q315Q XP_011540925.1

View Full Product Details