Product Details

SNP ID
rs202161857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78173855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAAGAATCGATGCAGAAGCCAT[C/T]GCTCAGAGCGATTTTCAGTACCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD21 PubMed Links

Gene Details

Gene
KCTD21
Gene Name
potassium channel tetramerization domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029859.2 786 Missense Mutation AAT,GAT N234D NP_001025030.1
XM_005273925.3 786 Missense Mutation AAT,GAT N303D XP_005273982.1
XM_005273928.1 786 Missense Mutation AAT,GAT N234D XP_005273985.1
XM_006718516.3 786 Missense Mutation AAT,GAT N270D XP_006718579.1
XM_006718517.2 786 Missense Mutation AAT,GAT N234D XP_006718580.1
XM_006718518.3 786 Missense Mutation AAT,GAT N234D XP_006718581.1
XM_011544955.2 786 Missense Mutation AAT,GAT N234D XP_011543257.1
Gene
KCTD21-AS1
Gene Name
KCTD21 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details