Product Details

SNP ID
rs202006620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102864213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGAAGTTCTTGGTAATCAGTTT[G/T]GGATAACCAGGGTCCATCATCTGTC
Phenotype
MIM: 601046
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MMP12 PubMed Links

Gene Details

Gene
MMP12
Gene Name
matrix metallopeptidase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002426.5 1342 Silent Mutation CCA,CCC P415P NP_002417.2

View Full Product Details