Product Details

SNP ID
rs201822633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:556547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCTGTGCAGGGCTGGGGCGACA[A/G]GACCGGCTCGCCGTGCCTGGGTGAG
Phenotype
MIM: 143023
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMNTD2 PubMed Links

Gene Details

Gene
LMNTD2
Gene Name
lamin tail domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173573.2 1097 Silent Mutation CTG,TTG L340L NP_775844.2
XM_011519965.2 1097 Silent Mutation CTG,TTG L413L XP_011518267.2
XM_011519967.2 1097 Silent Mutation CTG,TTG L413L XP_011518269.2
XM_017017476.1 1097 Silent Mutation CTG,TTG L413L XP_016872965.1
XM_017017477.1 1097 Silent Mutation CTG,TTG L408L XP_016872966.1
XM_017017478.1 1097 Silent Mutation CTG,TTG L345L XP_016872967.1
XM_017017479.1 1097 Silent Mutation CTG,TTG L413L XP_016872968.1
Gene
LRRC56
Gene Name
leucine rich repeat containing 56
There are no transcripts associated with this gene.

Gene
MIR210HG
Gene Name
MIR210 host gene
There are no transcripts associated with this gene.

Gene
RASSF7
Gene Name
Ras association domain family member 7
There are no transcripts associated with this gene.

View Full Product Details