Product Details

SNP ID
rs201403683
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74493307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGGGGCCGCGCGCGGGCGTCCT[C/G]CAGGCCCTGGAGCTCGCACAGGCGC
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LIPT2 PubMed Links

Gene Details

Gene
LIPT2
Gene Name
lipoyl(octanoyl) transferase 2 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011545021.2 666 Missense Mutation CAG,GAG Q215E XP_011543323.2
Gene
LOC100287896
Gene Name
uncharacterized LOC100287896
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319240.1 666 Intron NP_001306169.1
XM_017017055.1 666 Intron XP_016872544.1

View Full Product Details